Gene Transcripts

Gene Symbol
Rbsn
Entrez Gene ID
78287
Gene Name
rabenosyn, RAB effector
Gene Aliases
5330426D11Rik, Zfyve20
Chromosome Location
Chr. 6: 92186712 - 92214901 on Build GRCm38
UniGene ID
Mm.290734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030081.2 NP_084357.2 12 3153
GenBank mRNA AK146277.1 - 1 207
BC049106.1 - 12 3153 6412839

View Full Product Details