Gene Transcripts
- Gene Symbol
- Lsm2
- Entrez Gene ID
- 27756
- Gene Name
- LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)
- Gene Aliases
- D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP
- Chromosome Location
- Chr. 17: 34981854 - 34985891 on Build GRCm38
- UniGene ID
- Mm.165735
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204273.1 |
NP_001191202.1
|
1, 2 |
245 |
|
NM_030597.3 |
NP_085100.1
|
1, 2 |
245 |
|
GenBank mRNA |
AF204146.1 |
- |
1, 2 |
216 |
|
AF204147.1 |
- |
1, 2 |
216 |
|
AF204148.1 |
- |
1, 2 |
216 |
|
AF204149.1 |
- |
1, 2 |
216 |
|
AF204150.1 |
- |
1, 2 |
216 |
|
AF204151.1 |
- |
1, 2 |
216 |
|
AF204152.1 |
- |
1, 2 |
216 |
|
AF204154.1 |
- |
1, 2 |
216 |
|
AF204155.1 |
- |
1, 2 |
216 |
|
AF204156.1 |
- |
1, 2 |
166 |
|
BB848823.1 |
- |
1, 2 |
245 |
|
BC049543.1 |
- |
|
238 |
6774564 |
View Full Product Details