Gene Transcripts

Gene Symbol
Hspb8
Entrez Gene ID
80888
Gene Name
heat shock protein 8
Gene Aliases
AU018630, AW413033, Cryac, D5Ucla4, E2IG1, H11, H11K, HSP20-like, HSP22
Chromosome Location
Chr. 5: 116408495 - 116422864 on Build GRCm38
UniGene ID
Mm.21549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF250139.1 - 1, 2 392
AF273453.1 - 1, 2 738
AK005052.1 - 1, 2 760
BC003971.1 - 1, 2 748
BC011219.1 - 1, 2 741 4195285

View Full Product Details