Gene Transcripts
- Gene Symbol
- Hist1h1a
- Entrez Gene ID
- 80838
- Gene Name
- histone cluster 1, H1a
- Gene Aliases
- H1.1, H1a, H1f1, H1var3
- Chromosome Location
- Chr. 13: 23763668 - 23764412 on Build GRCm38
- UniGene ID
- Mm.480242
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC116820.1 |
- |
1 |
684 |
|
BC116850.1 |
- |
1 |
682 |
40126169 |
View Full Product Details