Gene Transcripts

Gene Symbol
Rad54l2
Entrez Gene ID
81000
Gene Name
RAD54 like 2 (S. cerevisiae)
Gene Aliases
Arip4, D130058C05, G630026H09Rik, Srisnf2l
Chromosome Location
Chr. 9: 106688080 - 106789213 on Build GRCm38
UniGene ID
Mm.246689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030730.2 NP_109655.2 13 2412
GenBank mRNA AJ132389.1 - 13 2313
AK083903.1 - 13 2393
AK143506.1 - 12 2028
AK155151.1 - 13 2369
BC133714.1 - 13 2261 40143565

View Full Product Details