Gene Transcripts

Gene Symbol
Tex15
Entrez Gene ID
104271
Gene Name
testis expressed gene 15
Gene Aliases
2210014E14Rik, AL022622, AU022940
Chromosome Location
Chr. 8: 33516684 - 33585585 on Build GRCm38
UniGene ID
Mm.280624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031374.2 NP_113551.2 1 149
GenBank mRNA AF285589.1 - 1 149
AK143304.1 - 6 921

View Full Product Details