Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031388.2 | NP_113565.2 | 8 | 1230 | |
GenBank mRNA | AF285570.1 | - | 5 | 826 | |
AK082829.1 | - | 8 | 1158 | ||
AK133417.1 | - | 8 | 1234 | ||
BC125454.1 | - | 3 | 729 | 40129969 | |
BC132154.1 | - | 3 | 729 | 40130431 |