Gene Transcripts
- Gene Symbol
- Mbd6
- Entrez Gene ID
- 110962
- Gene Name
- methyl-CpG binding domain protein 6
- Gene Aliases
- D10Wsu93e
- Chromosome Location
- Chr. 10: 127281956 - 127290719 on Build GRCm38
- UniGene ID
- Mm.372826
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033072.2 |
NP_149063.2
|
11, 12 |
3026 |
|
GenBank mRNA |
AK018262.1 |
- |
2, 3 |
167 |
|
AK047331.1 |
- |
1, 2 |
51 |
|
AK158797.1 |
- |
11, 12 |
3144 |
|
BC019433.1 |
- |
3, 4 |
527 |
|
BC038259.1 |
- |
2, 3 |
203 |
|
BC049833.1 |
- |
3, 4 |
510 |
|
BC055445.1 |
- |
3, 4 |
483 |
|
BC083328.1 |
- |
2, 3 |
163 |
|
BC096027.1 |
- |
8, 9 |
2768 |
|
View Full Product Details