Gene Transcripts

Gene Symbol
Slc4a10
Entrez Gene ID
94229
Gene Name
solute carrier family 4, sodium bicarbonate cotransporter-like, member 10
Gene Aliases
NCBE, mKIAA4136
Chromosome Location
Chr. 2: 62046515 - 62326743 on Build GRCm38
UniGene ID
Mm.314497
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242378.1 NP_001229307.1 2, 3 268
NM_001242379.1 NP_001229308.1 2, 3 268
NM_001242380.1 NP_001229309.1 2, 3 268
NM_001242381.1 NP_001229310.1 2, 3 268
NM_001242382.1 NP_001229311.1 2, 3 268
NM_001242383.1 NP_001229312.1 2, 3 268
NM_033552.3 NP_291030.2 2, 3 268
GenBank mRNA AB033759.1 - 2, 3 189
AK043017.1 - 2, 3 244
AK220501.1 - 2, 3 233
BC039226.1 - 2, 3 220 4480841
CF746204.1 - 2, 3 268
HM149278.1 - 2, 3 229
HQ533147.1 - 2, 3 208
HQ825316.1 - 2, 3 208
HQ831517.1 - 2, 3 208
JF500487.1 - 2, 3 345
JF500488.1 - 2, 3 345
JF500489.1 - 2, 3 345
JF500490.1 - 2, 3 345
JX220977.1 - 2, 3 235
JX254917.1 - 2, 3 235
JX268544.1 - 2, 3 346

View Full Product Details