Gene Transcripts

Gene Symbol
Vps37a
Entrez Gene ID
52348
Gene Name
vacuolar protein sorting 37A (yeast)
Gene Aliases
2210018P21Rik, 4930592A21Rik, AW261445, D8Ertd531e
Chromosome Location
Chr. 8: 40511783 - 40551134 on Build GRCm38
UniGene ID
Mm.489696
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033560.3 NP_291038.2 12 3653
GenBank mRNA AK011921.1 - 1 690
BC039290.1 - 12 3627 5066508
BC089318.1 - 1 980

View Full Product Details