Gene Transcripts

Gene Symbol
Pcdhga6
Entrez Gene ID
93714
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
Chromosome Location
Chr. 18: 37707229 - 37841870 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033589.1 NP_291067.1 1 413
GenBank mRNA AY013800.1 - 1 413
BC059927.1 - 1 548
BC080852.1 - 1 458 6844625

View Full Product Details