Gene Transcripts

Gene Symbol
Tac4
Entrez Gene ID
93670
Gene Name
tachykinin 4
Gene Aliases
AW489379, PPT-C
Chromosome Location
Chr. 11: 95261529 - 95269262 on Build GRCm38
UniGene ID
Mm.40085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053093.1 NP_444323.1 1 248
GenBank mRNA AF235035.1 - 1 248
AF515827.1 - 1 82
BC119426.1 - 1 228 40039772

View Full Product Details