Gene Transcripts
- Gene Symbol
- Ear6
- Entrez Gene ID
- 93719
- Gene Name
- eosinophil-associated, ribonuclease A family, member 6
- Gene Aliases
- Chromosome Location
- Chr. 14: 51853768 - 51854643 on Build GRCm38
- UniGene ID
- Mm.221324
- Species
- Mus musculus
- Gene Symbol
- Ear7
- Entrez Gene ID
- 53873
- Gene Name
- eosinophil-associated, ribonuclease A family, member 7
- Gene Aliases
- mR7
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.221324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017385.1 |
NP_059081.1
|
1 |
164 |
|
View Full Product Details