Gene Transcripts

Gene Symbol
Immp2l
Entrez Gene ID
93757
Gene Name
IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae)
Gene Aliases
AI853880, IMP2
Chromosome Location
Chr. 12: 41024090 - 41133662 on Build GRCm38
UniGene ID
Mm.363813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053122.4 NP_444352.2 260
GenBank mRNA AF359564.1 - 192
AK053361.1 - 234
AK161351.1 - 400
BC061049.1 - 260
BC100557.1 - 212 5057188

View Full Product Details