Gene Transcripts

Gene Symbol
Pcdhga7
Entrez Gene ID
93715
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
Chromosome Location
Chr. 18: 37714834 - 37841873 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033590.3 NP_291068.2 1 1038
GenBank mRNA AK034910.1 - 1 1055
AY013801.1 - 1 930
BC077712.1 - 1 1038 6825399

View Full Product Details