Gene Transcripts

Gene Symbol
Pcdhb6
Entrez Gene ID
93877
Gene Name
protocadherin beta 6
Gene Aliases
Pcdhb5B, PcdhbF
Chromosome Location
Chr. 18: 37334028 - 37336346 on Build GRCm38
UniGene ID
Mm.450193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053131.1 NP_444361.1 1 2051
GenBank mRNA AK032193.1 - 1664
AY013788.1 - 1 2051

View Full Product Details