Gene Transcripts

Gene Symbol
Pcdhb8
Entrez Gene ID
93879
Gene Name
protocadherin beta 8
Gene Aliases
Pcdhb5C, PcdhbH
Chromosome Location
Chr. 18: 37355271 - 37357610 on Build GRCm38
UniGene ID
Mm.348052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053133.1 NP_444363.1 1 252
GenBank mRNA AY013790.1 - 1 252
BC116268.1 - 1 253
BC116269.1 - 1 245
BC127160.1 - 1 252 40094058

View Full Product Details