Gene Transcripts

Gene Symbol
Mrpl43
Entrez Gene ID
94067
Gene Name
mitochondrial ribosomal protein L43
Gene Aliases
4930442D21Rik, bMRP36a
Chromosome Location
Chr. 19: 45005014 - 45006442 on Build GRCm38
UniGene ID
Mm.44174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053164.3 NP_444394.1 3 828
GenBank mRNA AA253916.1 - 1 90
BC065812.1 - 3 775

View Full Product Details