Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039551.1 | NP_001034640.1 | 3 | 1441 | |
NM_053186.2 | NP_444416.2 | 3 | 1441 | ||
GenBank mRNA | AF216964.1 | - | 3 | 510 | |
AK143035.1 | - | 2 | 143 | ||
BC052714.1 | - | 3 | 508 | ||
BC080275.1 | - | 3 | 868 | ||
BC108417.1 | - | 3 | 695 | ||
BC132285.1 | - | 3 | 493 | ||
BC144720.1 | - | 3 | 492 | ||
GQ225746.2 | - | 3 | 427 |