Gene Transcripts

Gene Symbol
Vmn1r50
Entrez Gene ID
113852
Gene Name
vomeronasal 1 receptor 50
Gene Aliases
V1RA5, V1rb1, VN2
Chromosome Location
Chr. 6: 90107275 - 90108207 on Build GRCm38
UniGene ID
Mm.377149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053225.1 NP_444455.1 1 790
GenBank mRNA BC107183.2 - 1 790
BC115706.1 - 1 789

View Full Product Details