Gene Transcripts

Gene Symbol
Ugt2a1
Entrez Gene ID
94215
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide A1
Gene Aliases
Chromosome Location
Chr. 5: 87459490 - 87490871 on Build GRCm38
UniGene ID
Mm.26794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053184.2 NP_444414.2 2 385
GenBank mRNA AF184901.1 - 1 301
AK140757.1 - 2 386
BC048926.1 - 1 373 6314878

View Full Product Details