Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000450.2 | NP_000441.2 | 14 | 2253 | |
GenBank mRNA | BC142677.1 | - | 14 | 2240 | |
BC142711.1 | - | 14 | 2240 | 40148829 | |
M24736.1 | - | 14 | 2212 | ||
M30640.1 | - | 14 | 2236 |