Gene Transcripts

Gene Symbol
Rasa2
Entrez Gene ID
114713
Gene Name
RAS p21 protein activator 2
Gene Aliases
5430433H21Rik, AA517451, AU023900, GAP1m
Chromosome Location
Chr. 9: 96539300 - 96631615 on Build GRCm38
UniGene ID
Mm.124502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053268.2 NP_444498.2 2 225
GenBank mRNA AB056433.1 - 2 218
AK034434.1 - 2 226
AK154499.1 - 2 242
AK166654.1 - 2 225

View Full Product Details