Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083937.1 | NP_001077406.1 | 3, 4 | 533 | |
NM_078484.2 | NP_511039.2 | 3, 4 | 533 | ||
GenBank mRNA | AB027147.1 | - | 3, 4 | 452 | |
AF229634.1 | - | 3, 4 | 533 | ||
AK146830.1 | - | 3, 4 | 449 | ||
AK155755.1 | - | 3, 4 | 445 | ||
BC037701.1 | - | 3, 4 | 470 | 4187325 |