Gene Transcripts

Gene Symbol
Akr1c20
Entrez Gene ID
116852
Gene Name
aldo-keto reductase family 1, member C20
Gene Aliases
2610528B18Rik
Chromosome Location
Chr. 13: 4486972 - 4523354 on Build GRCm38
UniGene ID
Mm.37605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054080.1 NP_473421.1 2, 3 279
GenBank mRNA AB097220.1 - 2, 3 267
AK012162.1 - 2, 3 279
BC021607.1 - 2, 3 275 5098938

View Full Product Details