Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080854.3 | NP_543130.2 | 5 | 429 | |
GenBank mRNA | AB054999.1 | - | 5 | 409 | |
BC125326.1 | - | 4 | 362 | 40129841 | |
BC131997.1 | - | 4 | 362 | 40130274 | |
CJ068489.1 | - | 5 | 423 |