Gene Transcripts

Gene Symbol
Krtap19-2
Entrez Gene ID
170651
Gene Name
keratin associated protein 19-2
Gene Aliases
Krtap16-1
Chromosome Location
Chr. 16: 88873665 - 88874269 on Build GRCm38
UniGene ID
Mm.196827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130870.1 NP_570940.1 1 512
GenBank mRNA AF345291.1 - 1 512
AK137740.1 - 1 208

View Full Product Details