Gene Transcripts

Gene Symbol
Eps8l2
Entrez Gene ID
98845
Gene Name
EPS8-like 2
Gene Aliases
AI042819, AW545405, Eps8r2
Chromosome Location
Chr. 7: 141338911 - 141363016 on Build GRCm38
UniGene ID
Mm.27451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133191.2 NP_573454.2 4 210
GenBank mRNA BC005492.1 - 4 210
BC009098.1 - 4 268 3482190

View Full Product Details