Gene Transcripts

Gene Symbol
Dazap1
Entrez Gene ID
70248
Gene Name
DAZ associated protein 1
Gene Aliases
2410042M16Rik, mPrrp
Chromosome Location
Chr. 10: 80261471 - 80288413 on Build GRCm38
UniGene ID
Mm.148693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122604.1 NP_001116076.1 6 590
NM_001122605.1 NP_001116077.1 6 587
NM_133188.2 NP_573451.2 6 587
GenBank mRNA AF225910.1 - 6 481
AK148023.1 - 6 465
AK152314.1 - 6 557
AK152345.1 - 6 557
BC049355.1 - 6 507 6337388
BC058775.1 - 6 501

View Full Product Details