Gene Transcripts

Gene Symbol
Cyp2d13
Entrez Gene ID
68444
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 13
Gene Aliases
1300007K12Rik
Chromosome Location
Chr. 15: 82636750 - 82642045 on Build GRCm38
UniGene ID
Mm.443212
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004933.1 - 271
BC018344.1 - 2, 3 421
BY705142.1 - 1, 2 269

View Full Product Details