Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205231.1 | NP_001192160.1 | 7 | 836 | |
NM_133671.3 | NP_598432.2 | 7 | 836 | ||
GenBank mRNA | AK078496.1 | - | 7 | 790 | |
BC007487.1 | - | 3 | 1070 | 3593924 | |
BC043071.1 | - | 7 | 836 | 6403830 | |
BC094451.1 | - | 5 | 489 | ||
BC106134.1 | - | 6 | 552 | 6309073 | |
X64587.1 | - | 7 | 742 |