Gene Transcripts

Gene Symbol
Lyrm5
Entrez Gene ID
67636
Gene Name
LYR motif containing 5
Gene Aliases
4930469P12Rik, AL022899, Ghiso
Chromosome Location
Chr. 6: 145211134 - 145216542 on Build GRCm38
UniGene ID
Mm.486359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163628.1 NP_001157100.1 3 563
NM_133688.3 NP_598449.1 3 326
GenBank mRNA AF412298.1 - 3 980
AF412299.1 - 4 561
AF412300.1 - 3 327
AK015530.1 - 3 299
AK075835.1 - 3 307
AY753325.1 - 3 318
BC021522.1 - 3 274 5352723
CX203242.1 - 3 563

View Full Product Details