Gene Transcripts

Gene Symbol
Bloc1s4
Entrez Gene ID
117197
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino
Gene Aliases
2610101N07Rik, Blos4, Cno
Chromosome Location
Chr. 5: 36747374 - 36748679 on Build GRCm38
UniGene ID
Mm.46744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133724.3 NP_598485.1 1 740
GenBank mRNA AK011795.1 - 1 712
BC019169.1 - 1 730 5002867

View Full Product Details