Gene Transcripts

Gene Symbol
Ube2s
Entrez Gene ID
77891
Gene Name
ubiquitin-conjugating enzyme E2S
Gene Aliases
0910001J09Rik, 6720465F12Rik, AA409170, E2-EPF
Chromosome Location
Chr. 7: 4808014 - 4812340 on Build GRCm38
UniGene ID
Mm.389787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133777.2 NP_598538.1 3 411
GenBank mRNA AK003078.1 - 3 425
AK017648.1 - 3 427
AK020143.1 - 423
AK146276.1 - 3 423
AK154067.1 - 3 427
BC012255.1 - 3 411 3581845
BC030171.1 - 3 405 5037250
BC083323.1 - 3 393 5715696

View Full Product Details