Gene Transcripts

Gene Symbol
Nol12
Entrez Gene ID
97961
Gene Name
nucleolar protein 12
Gene Aliases
C78541, Nop25
Chromosome Location
Chr. 15: 78934933 - 78941910 on Build GRCm38
UniGene ID
Mm.157216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133800.3 NP_598561.2 6 1065
GenBank mRNA AK036891.1 - 6 1008
AK037035.1 - 6 1059
AK083539.1 - 1067

View Full Product Details