Gene Transcripts

Gene Symbol
Chmp7
Entrez Gene ID
105513
Gene Name
charged multivesicular body protein 7
Gene Aliases
4930596K11Rik, 6330407G04Rik, AI450338, AW550775
Chromosome Location
Chr. 14: 69716979 - 69732570 on Build GRCm38
UniGene ID
Mm.159637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134078.4 NP_598839.2 1 173
GenBank mRNA BC024115.1 - 1 161
BC033365.1 - 1 144 5367653
BY129213.1 - 1 174

View Full Product Details