Gene Transcripts
- Gene Symbol
- Cyp2c50
- Entrez Gene ID
- 107141
- Gene Name
- cytochrome P450, family 2, subfamily c, polypeptide 50
- Gene Aliases
- AI266900
- Chromosome Location
- Chr. 19: 40089679 - 40113955 on Build GRCm38
- UniGene ID
- Mm.474575
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC011222.1 |
- |
5, 6 |
831 |
|
BC051050.1 |
- |
4, 5 |
662 |
5052536 |
View Full Product Details