Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134077.4 | NP_598838.3 | 11 | 2055 | |
GenBank mRNA | AK049083.1 | - | 10 | 1510 | |
AK051686.1 | - | 11 | 2031 | ||
AK139077.1 | - | 11 | 2056 | ||
AK172039.1 | - | 11 | 2034 | ||
BC049360.1 | - | 11 | 2032 | ||
BC066051.1 | - | 11 | 2007 |