Gene Transcripts
- Gene Symbol
- Vmn1r80
- Entrez Gene ID
- 171238
- Gene Name
- vomeronasal 1 receptor 80
- Gene Aliases
- V1rg3
- Chromosome Location
- Chr. 7: 12192965 - 12193891 on Build GRCm38
- UniGene ID
- Mm.438893
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134204.2 |
NP_598965.2
|
1 |
462 |
|
View Full Product Details