Gene Transcripts

Gene Symbol
Tango2
Entrez Gene ID
27883
Gene Name
transport and golgi organization 2
Gene Aliases
D16H22S680E, T10
Chromosome Location
Chr. 16: 18300825 - 18348098 on Build GRCm38
UniGene ID
Mm.265990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138583.2 NP_613049.2 6 475
GenBank mRNA AK089239.1 - 6 457
AK133588.1 - 6 458
AK168595.1 - 5 454
AK199175.1 - 1 17
BC005445.1 - 6 536 3496749
X74504.1 - 5 448

View Full Product Details