Gene Transcripts

Gene Symbol
Tbc1d10b
Entrez Gene ID
68449
Gene Name
TBC1 domain family, member 10b
Gene Aliases
1110003P22Rik, C87963
Chromosome Location
Chr. 7: 127197459 - 127208468 on Build GRCm38
UniGene ID
Mm.41420
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144522.5 NP_653105.3 5 1370
GenBank mRNA AF285112.1 - 5 728
AK047320.1 - 5 1371
AK047327.1 - 5 1371
BC072576.1 - 5 913 6401332

View Full Product Details