Gene Transcripts

Gene Symbol
Cdc26
Entrez Gene ID
66440
Gene Name
cell division cycle 26
Gene Aliases
2010012C09Rik
Chromosome Location
Chr. 4: 62393812 - 62408640 on Build GRCm38
UniGene ID
Mm.109930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139291.3 NP_647452.1 4 598
GenBank mRNA AK004518.1 - 1 238
AK012627.1 - 4 599
AK047055.1 - 618
AK086692.1 - 1077
AK135556.1 - 598
BC005775.1 - 3 525 3602527
BC114350.2 - 2 385 40053418

View Full Product Details