Gene Transcripts

Gene Symbol
Spice1
Entrez Gene ID
212514
Gene Name
spindle and centriole associated protein 1
Gene Aliases
Ccdc52, D16Ertd480e
Chromosome Location
Chr. 16: 44347395 - 44388492 on Build GRCm38
UniGene ID
Mm.24035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144550.4 NP_653133.3 8 747
GenBank mRNA AK048789.1 - 8 691
AK165031.1 - 6 590
AY099108.1 - 8 1027
BC052381.1 - 8 701 5703283
BC056960.1 - 8 701 5703283

View Full Product Details