Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144836.2 | NP_659085.1 | 9 | 1431 | |
GenBank mRNA | AK040717.1 | - | 9 | 1432 | |
AK050184.1 | - | 8 | 1867 | ||
AK050277.1 | - | 8 | 1761 | ||
BC018306.1 | - | 8 | 1069 | 4193755 |