Gene Transcripts

Gene Symbol
Tifa
Entrez Gene ID
211550
Gene Name
TRAF-interacting protein with forkhead-associated domain
Gene Aliases
T2bp
Chromosome Location
Chr. 3: 127788857 - 127798394 on Build GRCm38
UniGene ID
Mm.31852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145133.3 NP_660115.1 3 529
GenBank mRNA AB062111.1 - 3 326
AK041891.1 - 2 363
AK044221.1 - 2 473
AK149408.1 - 3 365
AK149864.1 - 3 497
AK151651.1 - 3 480
AK166239.1 - 3 571
BC065775.1 - 3 524 1528121

View Full Product Details