Gene Transcripts

Gene Symbol
Pomt1
Entrez Gene ID
99011
Gene Name
protein-O-mannosyltransferase 1
Gene Aliases
AI505244
Chromosome Location
Chr. 2: 32236595 - 32255005 on Build GRCm38
UniGene ID
Mm.31192
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145145.1 NP_660127.1 11 1120
GenBank mRNA AK053889.1 - 10 1640
AK134770.1 - 11 1164
AK153984.1 - 11 1219
AY007238.1 - 11 1160
AY494857.1 - 11 1180
BC027325.1 - 11 1120 3981366

View Full Product Details