Gene Transcripts
- Gene Symbol
- Frmd4b
- Entrez Gene ID
- 232288
- Gene Name
- FERM domain containing 4B
- Gene Aliases
- 6030440G05Rik, C87375, GOBLIN, GRSP1, R74720, mKIAA1013
- Chromosome Location
- Chr. 6: 97286867 - 97617657 on Build GRCm38
- UniGene ID
- Mm.27789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145148.2 |
NP_660130.2
|
13, 14 |
1389 |
|
GenBank mRNA |
AB042027.1 |
- |
13, 14 |
1312 |
|
AF327856.1 |
- |
12, 13 |
1012 |
|
AF327857.1 |
- |
11, 12 |
799 |
|
AK031491.1 |
- |
2, 3 |
575 |
|
AK129262.1 |
- |
12, 13 |
864 |
|
AK132280.1 |
- |
13, 14 |
1253 |
|
AK167170.1 |
- |
1, 2 |
1777 |
|
BC052390.1 |
- |
13, 14 |
1258 |
5718942 |
BC058262.1 |
- |
2, 3 |
212 |
6487449 |
View Full Product Details