Gene Transcripts

Gene Symbol
Alg1
Entrez Gene ID
208211
Gene Name
asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase)
Gene Aliases
HMAT1, HMT1
Chromosome Location
Chr. 16: 5233621 - 5244907 on Build GRCm38
UniGene ID
Mm.34581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145362.2 NP_663337.2 3 355
GenBank mRNA AK145871.1 - 3 355
BC011281.1 - 2 262

View Full Product Details