Gene Transcripts
- Gene Symbol
- Appl2
- Entrez Gene ID
- 216190
- Gene Name
- adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
- Gene Aliases
- Dip3b
- Chromosome Location
- Chr. 10: 83600033 - 83648854 on Build GRCm38
- UniGene ID
- Mm.282985
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145220.2 |
NP_660255.1
|
5, 6 |
454 |
|
GenBank mRNA |
AK154525.1 |
- |
5, 6 |
478 |
|
AK160075.1 |
- |
5, 6 |
433 |
|
AK160105.1 |
- |
5, 6 |
430 |
|
AK161003.1 |
- |
5, 6 |
461 |
|
AK168403.1 |
- |
5, 6 |
415 |
|
AK168748.1 |
- |
5, 6 |
410 |
|
AK171512.1 |
- |
5, 6 |
410 |
|
AY113706.1 |
- |
5, 6 |
456 |
|
BC048906.1 |
- |
5, 6 |
392 |
6475576 |
View Full Product Details