Gene Transcripts

Gene Symbol
Wfdc5
Entrez Gene ID
209232
Gene Name
WAP four-disulfide core domain 5
Gene Aliases
BC019734, Prg5, Wap1
Chromosome Location
Chr. 2: 164176325 - 164182742 on Build GRCm38
UniGene ID
Mm.218273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145369.3 NP_663344.2 4 685
GenBank mRNA AK075896.1 - 3 375
AK131906.1 - 4 618
AY669505.1 - 4 684
BC019734.1 - 4 685 3987617

View Full Product Details